missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Bovine Growth Serum
A cost-effective alternative to Fetal Bovine Serum (FBS).
Brand: Cytiva SH30541.02HI
55.72 GBP valid until 2024-12-31
Use promo code "21631" to get your promotional price.
This item is not returnable.
View return policy
Description
Specifications
Insulin, Gamma Globulin, Alkaline Phosphatase, Lactate Dehydrogenase, SGPT, SGOT, pH, Total Protein, Albumin, Blood Urea Nitrogen, Creatinine, Total Bilirubin, Sodium, Potassium, Calcium, Chloride, Phosphorus, Osmolality, Iron, TIBC, Percent Saturation, Glucose, IgG | |
Heat Inact.; 100 mL; Available as quote | |
100 mL | |
Sequential-filtered, 100 nm | |
Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus | |
Yes | |
Yes |
Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C. | |
Bovine Growth Serum | |
Sterile | |
Heat Inactivated | |
≤ 10 EU/mL | |
≤ 10 mg/dL |