missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Bovine Growth Serum
A cost-effective alternative to Fetal Bovine Serum (FBS).
Brand: Cytiva SH30541.03IR
Code : 85
Additional Details : Weight : 0.01000kg
Specifications
Insulin, Gamma Globulin, Alkaline Phosphatase, Lactate Dehydrogenase, SGPT, SGOT, pH, Total Protein, Albumin, Blood Urea Nitrogen, Creatinine, Total Bilirubin, Sodium, Potassium, Calcium, Chloride, Phosphorus, Osmolality, Iron, TIBC, Percent Saturation, Glucose, IgG | |
Gamma Irradiated; 500 mL; Available as quote | |
500 mL | |
Sequential-filtered, 100 nm | |
Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus | |
≤ 10 mg/dL |
Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C. | |
Bovine Growth Serum | |
Sterile | |
Irradiated | |
≤ 10 EU/mL | |
Yes |