missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ Bovine Growth Serum

A cost-effective alternative to Fetal Bovine Serum (FBS).

Brand:  Cytiva SH30541.03IR25-40

220.40 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.



Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 16201122

  • £232.00 / 500mL
Estimated Shipment: 04-09-2024
to see stock.

Description

Description

Specifications

Specifications

Insulin, Gamma Globulin, Alkaline Phosphatase, Lactate Dehydrogenase, SGPT, SGOT, pH, Total Protein, Albumin, Blood Urea Nitrogen, Creatinine, Total Bilirubin, Sodium, Potassium, Calcium, Chloride, Phosphorus, Osmolality, Iron, TIBC, Percent Saturation, Glucose, IgG
500 mL
Sequential-filtered, 100 nm
Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus
No
Yes
<–1°C
Sterile
Irradiated (25 to 40 doses)
≤ 10 EU/mL
≤ 10 mg/dL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers