Learn More
Cytiva HyClone™ Calf Serum (U.S.)
A cost-effective alternative to Fetal Bovine Serum (FBS).
Brand: Cytiva SH30073.04
103.52 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Code : 85
Additional Details : Weight : 1.00000kg
Description
Supplement cell culture growth with U.S.-sourced HyClone™ Bovine Calf Serum, an excellent and cost-effective alternative to fetal bovine serum (FBS).
HyClone Bovine Calf Serum is available with or without iron supplementation. Bovine calf serum products contain exceptionally high levels of transferrin, which, when supplemented, can provide three to four times as much available iron as FBS. In many applications, the performance of calf serum will equal or even surpass that of FBS. Bovine Calf Serum is filtered through three sequential 100 nm (0.1 μm) pore size-rated filters. Before dispensing, each lot of serum is pooled using true pool technology to ensure uniformity and consistency between bottles.
Specifications
Albumin, Alkaline Phosphatase, Lactate Dehydrogenase, Calcium, Percent Saturation, pH, Gamma Globulin, Potassium, IgG, SGPT, Sodium, Alkaline Phosphatase, TIBC, Total Bilirubin, Total Protein, Blood Urea Nitrogen, Calcium, Chloride, Creatinine, Gamma Globulin, Glucose, IgG, Insulin | |
Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C. | |
Calf Bovine Serum | |
Sterile | |
Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus |
Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities | |
Calf Serum (U.S.) | |
1000 mL | |
Sequential-filtered, 100 nm | |
Yes |