Learn More
Cytiva HyClone™ Calf Serum (U.S.)
A cost-effective alternative to Fetal Bovine Serum (FBS).
£13.65 - £129.00
Specifications
Certifications/Compliance | Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities |
---|---|
Content And Storage | Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C. |
Sterility | Sterile |
Sterilization Method | Sequential-filtered, 100 nm |
USDA Safety Testing Included | Yes |
Product Code | Brand | Description | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Quantity | Price | Quantity & Availability | ||||
10014813
|
Cytiva
SH30073.04 |
Calf Serum (U.S.) | 1000 mL |
£110.00
1000mL |
Estimated Shipment: 20-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11571831
|
Cytiva
SH30073.03 |
Calf Serum (U.S.) | 500 mL |
£83.95
500mL |
Estimated Shipment: 07-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12369812
|
Cytiva
SH30073.02HI |
Heat Inactivated Calf Serum (U.S.) | 100 mL |
£37.25
100mL |
Estimated Shipment: 17-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12869447
|
Cytiva
SH30073.04HI |
Heat Inactivated Calf Serum (U.S.) | 1000 mL |
£129.00
1 L |
Estimated Shipment: 20-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10546863
|
Cytiva
SH30073.02 |
Calf Serum (U.S.) | 100 mL |
£13.65
100mL |
Estimated Shipment: 02-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
16241092
|
Cytiva
SH30073.03IR25-40 |
Irradiated calf serum (U.S.), 25 to 40 doses | 500 mL |
£75.20
500mL |
Estimated Shipment: 20-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15735533
|
Cytiva
SH30073.04IR |
Irradiated Calf Serum (U.S.) | 1000 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12399812
|
Cytiva
SH30073.03IR |
Irradiated Calf Serum (U.S.) | 500 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
Supplement cell culture growth with U.S.-sourced HyClone™ Bovine Calf Serum, an excellent and cost-effective alternative to fetal bovine serum (FBS).
HyClone Bovine Calf Serum is available with or without iron supplementation. Bovine calf serum products contain exceptionally high levels of transferrin, which, when supplemented, can provide three to four times as much available iron as FBS. In many applications, the performance of calf serum will equal or even surpass that of FBS. Bovine Calf Serum is filtered through three sequential 100 nm (0.1 μm) pore size-rated filters. Before dispensing, each lot of serum is pooled using true pool technology to ensure uniformity and consistency between bottles.
Specifications
Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities | |
Sterile | |
Yes |
Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C. | |
Sequential-filtered, 100 nm |