missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ CDM4CHO™ Media, Liquid
Support growth of CHO cell culture. HyClone™ CDM4CHO™ Media, Liquid is a serum-free, chemically defined media.
Brand: Cytiva SH30558.LS
Code : 83
Additional Details : Weight : 0.50000kg
Description
- Format: Liquid
Specifications
cGMP | |
CDM4CHO media | |
Sodium Bicarbonate, Poloxamer 188 | |
6 x 1000 mL |
2° to 8°C | |
Liquid | |
No Glutamine, No Phenol Red, No HEPES, No Protein |