missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ CDM4CHO™ Media, Liquid
Support growth of CHO cell culture. HyClone™ CDM4CHO™ Media, Liquid is a serum-free, chemically defined media.
£70.80 - £272.67
Product Code | Brand | Quantity | With Additives | Without Additives | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | With Additives | Without Additives | Price | Quantity & Availability | |||
10463522
|
Cytiva
SH30558.02 |
1000 mL | Sodium Bicarbonate, Poloxamer 188 | No Glutamine, No Phenol Red, No HEPES, No Protein |
£97.11
1000mL |
Estimated Shipment: 06-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10433142
|
Cytiva
SH30557.02 |
1000 mL bottle | Glutamine, Sodium Bicarbonate, Poloxamer 188 | No Phenol Red, No HEPES, No Protein |
£70.80
1000mL |
Estimated Shipment: 09-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10298023
|
Cytiva
SH30558.LS |
6 x 1000 mL | Sodium Bicarbonate, Poloxamer 188 | No Glutamine, No Phenol Red, No HEPES, No Protein |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Description
- Format: Liquid
Specifications
cGMP | |
CDM4CHO media | |
Liquid |