Learn More
Cytiva HyClone™ CDM4CHO™ Media, Liquid
Support growth of CHO cell culture. HyClone™ CDM4CHO™ Media, Liquid is a serum-free, chemically defined media.
Brand:  Cytiva SH30557.02
74.26 GBP valid until 2025-02-21
Use promo code "24115" to get your promotional price.
 View more versions of this product
Product Code. 10433142
  • £78.70 / 1000mL
Explore more special offers
Description
Description
  • Format: Liquid
TRUSTED_SUSTAINABILITY
Specifications
Specifications
cGMP
CDM4CHO media
CDM4CHO Liquid Medium
Glutamine, Sodium Bicarbonate, Poloxamer 188
1000 mL bottle
2° to 8°C, away from light
Liquid
0.2 μm Sterile-filtered
No Phenol Red, No HEPES, No Protein