missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Characterized Fetal Bovine Serum (U.S. Origin)
Enhance growth of cells. Tested for endotoxin, hemoglobin, and additional biochemical assays.
Brand: Cytiva SH30071.01
73.67 GBP valid until 2024-06-28
Use promo code "21691" to get your promotional price.
Code : 85
Additional Details : Weight : 0.01000kg
Description
Specifications
Lactate Dehydrogenase,SGPT,SGOT,pH,Total Protein,Albumin,Blood Urea Nitrogen,Creatinine,Total Bilirubin,Sodium,Potassium,Calcium,Chloride,Inorganic Phosphorus,Osmolality,Iron,TIBC,Percent Saturation,Glucose,IgG,Gamma Globulin | |
Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C. | |
50 mL | |
Sequential Triple-filtered, 100 nm | |
None | |
≤ 25 EU/mL | |
≤ 25 mg/dL | |
USDA Inspected |
Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities | |
United States | |
Sterile | |
Characterized | |
Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus | |
No | |
No |