missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Characterized Fetal Bovine Serum (U.S. Origin)
Enhance growth of cells. Tested for endotoxin, hemoglobin, and additional biochemical assays.
£83.45 - £101.00
Specifications
Additional Assays | Lactate Dehydrogenase,SGPT,SGOT,pH,Total Protein,Albumin,Blood Urea Nitrogen,Creatinine,Total Bilirubin,Sodium,Potassium,Calcium,Chloride,Inorganic Phosphorus,Osmolality,Iron,TIBC,Percent Saturation,Glucose,IgG,Gamma Globulin |
---|---|
Certifications/Compliance | Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities |
Content And Storage | Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C. |
Country of Origin | United States |
Quantity | 50 mL |
Product Code | Brand | Processing | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Processing | Price | Quantity & Availability | |||||
12604217
|
Cytiva
SH30071.01HI |
Heat Inactivated |
£101.00
50mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11596531
|
Cytiva
SH30071.01 |
None |
£83.45
50mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Specifications
Lactate Dehydrogenase,SGPT,SGOT,pH,Total Protein,Albumin,Blood Urea Nitrogen,Creatinine,Total Bilirubin,Sodium,Potassium,Calcium,Chloride,Inorganic Phosphorus,Osmolality,Iron,TIBC,Percent Saturation,Glucose,IgG,Gamma Globulin | |
Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C. | |
50 mL | |
Sequential Triple-filtered, 100 nm | |
Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus | |
≤ 25 mg/dL | |
USDA Inspected |
Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities | |
United States | |
Sterile | |
Characterized | |
≤ 25 EU/mL | |
No |