missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Defined Fetal Bovine Serum (FBS), US Origin
Widely used by cell culturists who have a concern for viral contaminants and require an extensive biochemical profile.
Brand: Cytiva SH30070.03HT
Additional Details : Weight : 0.59000kg
Description
- Assay results for over 50 components, with additional testing available
- Sourced and processed in the United States with complete traceability to origin
- Ultra-low endotoxin levels (≤ 10 EU/mL) for reduced research and production risk
- Serial 40 nm filter process removes up to 8 logs viral challenge
- Post-filtration treatments such as γ-irradiated and heat-inactivated options are also available
- Broad virus panel testing in accordance with 9 CFR 113.53
Specifications
United States | |
500 mL | |
Defined | |
Virus-Tested to 9 CFR Standard | |
≤ 10 EU/mL | |
HyClone™ |
Fetal Bovine Serum | |
Sequential-filtered, 40 nm | |
Heat Inactivated | |
500 mL | |
≤ 10 mg/dL |