missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Defined Fetal Bovine Serum (FBS), US Origin
Widely used by cell culturists who have a concern for viral contaminants and require an extensive biochemical profile.
£202.00 - £875.00
Specifications
Country of Origin | United States |
---|---|
Product Type | Fetal Bovine Serum |
Sterilization Method | Sequential-filtered, 40 nm |
Grade | Defined |
Tested For | Virus-Tested to 9 CFR Standard |
Product Code | Brand | Processing | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Processing | Quantity | Price | Quantity & Availability | ||||
16211082
|
Cytiva
SH30070.03IH25-40 |
Irradiated (25-40 kGy) | 500 mL |
£875.00
500mL |
Estimated Shipment: 08-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
16221082
|
Cytiva
SH30070.03IH30-45 |
Irradiated (30-45 kGy) | 500 mL |
£875.00
500mL |
Estimated Shipment: 08-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
16675725
|
Cytiva
SH30070.02HT |
Heat Inactivated | 100 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
16695725
|
Cytiva
SH30070.03HT |
Heat Inactivated | 500 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
16685725
|
Cytiva
SH30070.02IH |
Irradiated, Heat Inactivated | 100 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
16605735
|
Cytiva
SH30070.03MH |
Mesenchymal Stem Cell (MSC) Screen, Heat Inactivated | 100 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
16251082
|
Cytiva
SH30070.03TIR25-40 |
TET Screen, Irradiated (24-40 kGy) | 500 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
- Assay results for over 50 components, with additional testing available
- Sourced and processed in the United States with complete traceability to origin
- Ultra-low endotoxin levels (≤ 10 EU/mL) for reduced research and production risk
- Serial 40 nm filter process removes up to 8 logs viral challenge
- Post-filtration treatments such as γ-irradiated and heat-inactivated options are also available
- Broad virus panel testing in accordance with 9 CFR 113.53
Specifications
United States | |
Sequential-filtered, 40 nm | |
Virus-Tested to 9 CFR Standard | |
≤ 10 mg/dL |
Fetal Bovine Serum | |
Defined | |
≤ 10 EU/mL | |
HyClone™ |