Learn More
Cytiva Hyclone™ Donor Equine Serum
Carefully collected from selected U.S. donor herds that receive regular veterinary inspection and care.
Brand:  Cytiva SH30074.04IR
Additional Details : Weight : 1.15240kg
 View more versions of this product
Product Code. 17286936
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
Serum Collection:
  • From collection to final packaging
  • Every step is documented
  • Full traceability compliant with current Good Manufacturing Practices (cGMP)
Quick and Reliable Processing:
  • Extensive quality, performance, and analytical testing
  • Aseptically packaged in a Class 100 clean room condition
Quality Control Testing Standards:
  • Each lot of serum is tested to ensure quality and sterility
True Pool Processing:
  • Pooled after filtration and before final packaging
  • Consistent quality from the first bottle filled to the last
  • Increases consistency and minimizes variability with a lot of serum
Specifications
Specifications
Albumin
Liquid
1 L
Sequential Triple-filtered, 100 nm
BVDV, Rabies Virus, Reovirus, Cytopathogenic Agents
HyClone™
United States
Equine Serum
Sterile
Irradiated
No