missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Donor Equine Serum, US Origin
Used to enhance cell culture performance; a cost-efficient alternative to Fetal Bovine Serum (FBS).
Brand: Cytiva SH30074.04IR25-40
124.45 GBP valid until 2024-06-28
Use promo code "21691" to get your promotional price.
Additional Details : Weight : 1.00000kg
Description
- Carefully collected from animals fed an enriched diet to ensure proper nutrition
- To reduce lipid concentration, the equine herd is fasted and then the blood is collected in a special facility using aseptic techniques
- Coggins tests for equine infectious anemia are performed on each horse
Specifications
>6 months | |
Liquid | |
1000 mL | |
Irradiated (25-40 kGy) | |
No | |
HyClone™ |
United States | |
Equine Serum | |
Sequential Triple-filtered, 100 nm | |
≤ 10 EU/mL | |
≤ 10 mg/dL |