missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Hyclone™ Characterized Fetal Bovine Serum (Australia)

Characterized FBS irradiated is comparable to the highest quality FBS from other suppliers, but is lower priced than defined FBS.

Brand:  Cytiva SH30084.04IR25-35

 View more versions of this product

Product Code. 17206946

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Filtered through 3 sequential 0.1 μm pore size filters
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Alkaline Phosphate, Lactate Dehydrogenase, SGPT, SGOT, Total Protein, Albumin, Blood Urea Nitrogen, Creatinine, Total Bilirubin, Sodium, Potassium, Calcium, Chloride, Inorganic Phosphorus, Glucose, Iron, TIBC, Percent Saturation, Gamma Globulin, IgG, pH, Osmolality
Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2 to 8°C.
Liquid
1 L
Sequential Triple-filtered, 100 nm, True Pool Technology
Irradiated (25 to 35 doses)
No
USDA Inspected
AQIS
Australia
Fetal Bovine Serum
Sterile
Characterized
Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus
HyClone™
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers