missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Hyclone™ Characterized Fetal Bovine Serum (Australia)

Characterized FBS irradiated is comparable to the highest quality FBS from other suppliers, but is lower priced than defined FBS.

£882.30 - £1723.80

Specifications

Certifications/Compliance AQIS
Content And Storage Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2 to 8°C.
Country of Origin Australia
Product Type Fetal Bovine Serum
Sterility Sterile
View More Specs

Products 8
Product Code Brand Quantity Processing Price Quantity & Availability  
Product Code Brand Quantity Processing Price Quantity & Availability  
12872926
View Documents
Cytiva
SH30084.03IH
500 mL Heat Inactivated This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11950958
View Documents
Cytiva
SH30084.03HI
500 mL Heat Inactivated
£1205.00
500mL
Please to purchase this item. Need a web account? Register with us today!
17266896
View Documents
Cytiva
SH30084.03IH25-40
500 mL Heat Inactivated, Irradiated (25 to 40 doses)
£1175.00
500mL
Please to purchase this item. Need a web account? Register with us today!
17276896
View Documents
Cytiva
SH30084.03IH30-45
500 mL Heat Inactivated, Irradiated (30 to 45 doses) This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16201102
View Documents
Cytiva
SH30084.03IR25-40
500 mL Irradiated (25 to 40 doses)
£1160.00
500mL
Please to purchase this item. Need a web account? Register with us today!
16221102
View Documents Promotion Details
Cytiva
500 mL Irradiated (30 to 45 doses)
£1160.00
500mL
Please to purchase this item. Need a web account? Register with us today!
17206946
View Documents
Cytiva
SH30084.04IR25-35
1 L Irradiated (25 to 35 doses) This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
17216946
View Documents
Cytiva
SH30084.04IR30-45
1 L Irradiated (30 to 45 doses) This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

  • Filtered through 3 sequential 0.1 μm pore size filters
Specifications

Specifications

AQIS
Australia
Sterile
USDA Inspected
Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2 to 8°C.
Fetal Bovine Serum
HyClone™
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.