missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Characterized Fetal Bovine Serum (New Zealand)
Enhance growth of cells with New Zealand sourced HyClone™ Characterized Fetal Bovine Serum.
Brand: Cytiva SH30406.03HI
Code : 85
Additional Details : Weight : 1.00000kg
Description
- HyClone Characterized Fetal Bovine Serum (FBS), New Zealand Origin is carefully collected, processed, and filtered in our own New Zealand facility.
- The great expanse of the Pacific ocean has protected New Zealand’s islands from many outside influences, both geographically and biologically.
- One of the benefits of this isolation is that New Zealand has the fewest reported bovine diseases in the world, making Characterized FBS, New Zealand Origin an excellent source of bovine serum as it offers greater security.
- Characterized FBS, New Zealand Origin is filtered through three sequential 100 nm (0.1 μm) pore size-rated filters.
- Before dispensing, each lot of serum is pooled to ensure uniformity and consistency between bottles (true pool technology).
Specifications
Progesterone, Chloride, Insulin, Inorganic Phosphorus, PTH, Glucose, Cholesterol, Copper, HDL, Magnesium, LDL, Selenium, Triglycerides, IgG, Gamma Globulin, pH, Phospholipids, Osmolality, Alkaline Phosphatase, Iron, Lactate Dehydrogenase, TIBC, SGPT, Percent Saturation, SGOT, B1, B2, B6, Niacin, Biotin, Retinol, Carotene, Tocopherol, B12, Folate, Vitamin C, 1,25 Dihydroxy Vitamin D, Total Protein, Albumin, and Blood Urea Nitrogen | |
New Zealand | |
Fetal Bovine Serum | |
Sterile | |
Characterized | |
Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus | |
Yes | |
HyClone™ |
Store at or below -10°C | |
Solution | |
1 L | |
Sequential-filtered, 100 nm | |
Heat Inactivated | |
≤ 20 EU/mL | |
≤ 25 mg/dL | |
USDA Inspected |