missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Characterized Fetal Bovine Serum (New Zealand)
Enhance growth of cells with New Zealand sourced HyClone™ Characterized Fetal Bovine Serum.
£985.00 - £1855.00
Specifications
Content And Storage | Store at or below -10°C |
---|---|
Country of Origin | New Zealand |
Product Type | Fetal Bovine Serum |
Sterility | Sterile |
Sterilization Method | Sequential-filtered, 100 nm |
Product Code | Brand | Quantity | Processing | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Processing | Price | Quantity & Availability | ||||
17256906
|
Cytiva
SH30406.02IR25-35 |
500 mL | Irradiated (25 to 35 doses) |
£985.00
500mL |
Estimated Shipment: 07-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12654217
|
Cytiva
SH30406.03 |
1 L | None |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12684227
|
Cytiva
SH30406.03HI |
1 L | Heat Inactivated |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12624247
|
Cytiva
SH30406.03IR |
1 L | Irradiated |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
17266906
|
Cytiva
SH30406.03IR30-40 |
1 L | Irradiated (30 to 40 doses) |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
- HyClone Characterized Fetal Bovine Serum (FBS), New Zealand Origin is carefully collected, processed, and filtered in our own New Zealand facility.
- The great expanse of the Pacific ocean has protected New Zealand’s islands from many outside influences, both geographically and biologically.
- One of the benefits of this isolation is that New Zealand has the fewest reported bovine diseases in the world, making Characterized FBS, New Zealand Origin an excellent source of bovine serum as it offers greater security.
- Characterized FBS, New Zealand Origin is filtered through three sequential 100 nm (0.1 μm) pore size-rated filters.
- Before dispensing, each lot of serum is pooled to ensure uniformity and consistency between bottles (true pool technology).
Specifications
Store at or below -10°C | |
Fetal Bovine Serum | |
Sequential-filtered, 100 nm | |
≤ 20 EU/mL | |
HyClone™ |
New Zealand | |
Sterile | |
Characterized | |
≤ 25 mg/dL | |
USDA Inspected |