missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Fetal Bovine Serum (U.S.), Charcoal/Dextran Treated
Augment growth of cells with U.S.-sourced HyClone™ Charcoal/Dextran Treated Fetal Bovine Serum (FBS), containing reduced levels of many hormones and growth factors.
Brand: Cytiva SH30068.02
143.45 GBP valid until 2024-12-31
Use promo code "21878" to get your promotional price.
This item is not returnable.
View return policy
Description
Specifications
1,Blood Urea Nitrogen,Calcitonin,Calcium,Carotene,Chloride,Corticosterone,Cortisol,Creatinine,Estrone,Folate,25 Dihydroxy Vitamin D,FSH,Glucose,IgG,Inorganic Phosphorus,Insulin,Lactate Dehydrogenase,LH,Niacin,Potassium,Progesterone,17 Beta Estradiol,Retinol,SGOT,SGPT,Sodium,T3,T4,Testosterone,Tocopherol,Total Bilirubin,Total Protein,Albumin,TSH,Vitamin C,Alkaline Phosphatase,B1,B12,B2,B6,Biotin | |
Charcoal/Dextran Treated Fetal Bovine Serum, US Origin, None | |
Charcoal/Dextran filtered | |
Sterile | |
Specialty filtered | |
Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus | |
No | |
No |
Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C | |
United States | |
100 mL | |
Sequential-filtered, 100 nm | |
None | |
≤ 10 EU/mL | |
≤ 20 mg/dL | |
USDA Inspected |
Safety and Handling
Hazard Statement
Supplemental information
- EUH210-Safety data sheet available on request.
Supplemental information
- UNKNOWN ACUTE-2.1 per cent of the mixture consists of component(s) of unknown acute toxicity.
- UNKNOWN AQUATIC-Contains 2.1% of components with unknown hazards to the aquatic environment.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction