missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Fetal Bovine Serum (U.S.), Charcoal/Dextran Treated
Augment growth of cells with U.S.-sourced HyClone™ Charcoal/Dextran Treated Fetal Bovine Serum (FBS), containing reduced levels of many hormones and growth factors.
Brand: Cytiva SH30068.02HI
157.70 GBP valid until 2024-12-31
Use promo code "21878" to get your promotional price.
This item is not returnable.
View return policy
Description
Specifications
1,Blood Urea Nitrogen,Calcitonin,Calcium,Carotene,Chloride,Corticosterone,Cortisol,Creatinine,Estrone,Folate,25 Dihydroxy Vitamin D,FSH,Glucose,IgG,Inorganic Phosphorus,Insulin,Lactate Dehydrogenase,LH,Niacin,Potassium,Progesterone,17 Beta Estradiol,Retinol,SGOT,SGPT,Sodium,T3,T4,Testosterone,Tocopherol,Total Bilirubin,Total Protein,Albumin,TSH,Vitamin C,Alkaline Phosphatase,B1,B12,B2,B6,Biotin | |
Charcoal/Dextran Treated Fetal Bovine Serum, US Origin, Heat Inactivated | |
Charcoal/Dextran filtered | |
100 mL | |
Sequential-filtered, 100 nm | |
Heat Inactivated | |
≤ 10 EU/mL | |
≤ 20 mg/dL | |
USDA Inspected |
Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C | |
United States | |
Fetal Bovine Serum | |
Sterile | |
Specialty filtered | |
Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus | |
Yes | |
No |
Safety and Handling
Hazard Statement
Supplemental information
- EUH210-Safety data sheet available on request.
Supplemental information
- UNKNOWN ACUTE-2.1 per cent of the mixture consists of component(s) of unknown acute toxicity.
- UNKNOWN AQUATIC-Contains 2.1% of components with unknown hazards to the aquatic environment.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction