missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ Fetal Bovine Serum (U.S.), Charcoal/Dextran Treated

Augment growth of cells with U.S.-sourced HyClone™ Charcoal/Dextran Treated Fetal Bovine Serum (FBS), containing reduced levels of many hormones and growth factors.

Brand:  Cytiva SH30068.01

84.12 GBP valid until 2024-12-31
Use promo code "21878" to get your promotional price.


 View more versions of this product

Product Code. 12389772

  • £93.50 / 50mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Processed using a proprietary charcoal/dextran treatment that has been shown to reduce the levels of many hormones and growth factors
  • This serum is assayed for a variety of components before and after treatment to ensure the efficacy of the treatment
  • Charcoal/dextran treatment has been shown to be effective in reducing steroid levels for use of the serum in receptor studies or estrogen-related investigations
  • Other studies indicate that charcoal/dextran treatments may be used to minimize lot-to-lot serum variability
  • Filtered through three sequential 100 nm (0.1 μm) pore size-rated filters
  • Before dispensing, each lot of serum is pooled using true pool technology to ensure uniformity and consistency between bottles
  • TRUSTED_SUSTAINABILITY
    Specifications

    Specifications

    1,Blood Urea Nitrogen,Calcitonin,Calcium,Carotene,Chloride,Corticosterone,Cortisol,Creatinine,Estrone,Folate,25 Dihydroxy Vitamin D,FSH,Glucose,IgG,Inorganic Phosphorus,Insulin,Lactate Dehydrogenase,LH,Niacin,Potassium,Progesterone,17 Beta Estradiol,Retinol,SGOT,SGPT,Sodium,T3,T4,Testosterone,Tocopherol,Total Bilirubin,Total Protein,Albumin,TSH,Vitamin C,Alkaline Phosphatase,B1,B12,B2,B6,Biotin
    Charcoal/Dextran Treated Fetal Bovine Serum, US Origin, None
    Charcoal/Dextran filtered
    Sterile
    Specialty filtered
    Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus
    No
    No
    Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C
    United States
    50 mL
    Sequential-filtered, 100 nm
    None
    ≤ 10 EU/mL
    ≤ 20 mg/dL
    USDA Inspected
    Product Suggestions

    Product Suggestions

    Videos
    Safety and Handling

    Safety and Handling

    Hazard Statement
    • EUH210-Safety data sheet available on request.

    Supplemental information
    • UNKNOWN ACUTE-2.1 per cent of the mixture consists of component(s) of unknown acute toxicity.
    • UNKNOWN AQUATIC-Contains 2.1% of components with unknown hazards to the aquatic environment.

    SDS
    Documents

    Documents

    Certificates
    Special Offers

    Special Offers

    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.