missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Fetal Bovine Serum (U.S.), Dialyzed
For applications that need serum without low molecular weight components.
Brand: Cytiva SH30079.03
572.21 GBP valid until 2024-12-31
Use promo code "21631" to get your promotional price.
This item is not returnable.
View return policy
Description
Augment growth of cells with U.S.-sourced HyClone™ Dialyzed Fetal Bovine Serum (FBS), depleted of small molecules (less than 10,000 MW) and filtered through sequential 100nm (0.1μm) pore-size rated filters.
Specifications
1, 25 Dihydroxy Vitamin D, Albumin, Alkaline Phosphatase, B1, B12, B2, B6, Biotin, Blood Urea Nitrogen, Calcium, Carotene, Chloride, Cholesterol, Copper, Creatinine, Folate, Gamma Globulin, Glucose, HDL, IgG, Inorganic Phosphorus, Insulin, Iron, Lactate Dehydrogenase, LDL, Magnesium, Niacin, Osmolality, Percent Saturation, pH, Phospholipids, Potassium, Progesterone, Retinol, Selenium, SGOT, SGPT, Sodium, TIBC, Tocopherol, Total Bilirubin, Total Protein, Triglycerides, Vitamin C | |
Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C. | |
United States | |
500 mL | |
Sequential-filtered, 100 nm | |
None | |
≤ 10 EU/mL | |
≤ 10 mg/dL | |
USDA Inspected |
EMEA Certified | |
Dialyzed Fetal Bovine Serum, US Origin, None | |
Dialyzed | |
Sterile | |
Specialty filtered | |
Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus | |
No | |
No |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction