missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Fetal Bovine Serum (U.S.), Dialyzed
For applications that need serum without low molecular weight components.
£86.80 - £675.00
Specifications
Country of Origin | United States |
---|---|
Specialty Type | Dialyzed |
Sterility | Sterile |
Sterilization Method | Sequential-filtered, 100 nm |
Grade | Specialty filtered |
Product Code | Brand | Quantity | Processing | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Processing | Price | Quantity & Availability | ||||
12349822
|
Cytiva
SH30079.01 |
50 mL | None |
£86.80
50mL |
Estimated Shipment: 06-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11521841
|
Cytiva
SH30079.02 |
100 mL | None |
£148.00
100mL |
Estimated Shipment: 06-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12379822
|
Cytiva
SH30079.02HI |
100 mL | Heat Inactivated |
£173.00
100mL |
Estimated Shipment: 03-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10594553
|
Cytiva
SH30079.03 |
500 mL | None |
£625.00
500mL |
Estimated Shipment: 09-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11960958
|
Cytiva
SH30079.03HI |
500 mL | Heat Inactivated |
£660.00
500mL |
Estimated Shipment: 19-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12604207
|
Cytiva
SH30079.01HI |
50 mL | Heat Inactivated |
£103.00
50mL |
Estimated Shipment: 19-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
16281092
|
Cytiva
SH30079.03IR25-40 |
500 mL | Gamma Irradiated (25 to 40 doses) |
£660.00
500mL |
Estimated Shipment: 08-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17652075
|
Cytiva
SH30079.03IH25-40 |
500 mL | Heat Inactivated, Gamma Irradiated (25 to 40 doses) |
£675.00
500mL |
Estimated Shipment: 05-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12309832
|
Cytiva
SH30079.03IR |
500 mL | Gamma Irradiated |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
Augment growth of cells with U.S.-sourced HyClone™ Dialyzed Fetal Bovine Serum (FBS), depleted of small molecules (less than 10,000 MW) and filtered through sequential 100nm (0.1μm) pore-size rated filters.
Specifications
United States | |
Sterile | |
Specialty filtered | |
≤ 10 mg/dL | |
USDA Inspected |
Dialyzed | |
Sequential-filtered, 100 nm | |
≤ 10 EU/mL | |
No |