Learn More
Cytiva HyClone™ Fetal Bovine Serum (U.S.), Dialyzed
For applications that need serum without low molecular weight components.
Brand: Cytiva SH30079.03IH25-40
610.35 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Additional Details : Weight : 0.59850kg
Description
Augment growth of cells with U.S.-sourced HyClone™ Dialyzed Fetal Bovine Serum (FBS), depleted of small molecules (less than 10,000 MW) and filtered through sequential 100nm (0.1μm) pore-size rated filters.
Specifications
25 Dihydroxy Vitamin D, Albumin, Alkaline Phosphatase, B1 | |
Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2°C to 8°C. | |
Dialyzed | |
Sterile | |
Specialty filtered | |
Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV | |
Yes | |
No |
EMEA Certified | |
United States | |
500 mL | |
Sequential-filtered, 100 nm | |
Heat Inactivated, Gamma Irradiated (25 to 40 doses) | |
≤ 10 EU/mL | |
≤ 10 mg/dL | |
USDA Inspected |