missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Fetal Bovine Serum (U.S.), Standard
Supplement cell culture growth with U.S. sourced quality cell culture-grade Cytiva HyClone™ Standard Fetal Bovine Serum (FBS)
£130.00 - £555.00
Specifications
Additional Assays | Gamma Globulin, Alkaline Phosphatase, Lactate Dehydrogenase, SGPT, SGOT, pH, Total Protein, Albumin, Blood Urea Nitrogen, Creatinine, Total Bilirubin, Sodium, Potassium, Calcium, Chloride, Inorganic Phosphorus, Osmolality, Iron, TIBC, IgG |
---|---|
Certifications/Compliance | Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities |
Country of Origin | United States |
Sterility | Sterile |
Sterilization Method | Sequential-filtered, 100 nm |
Product Code | Brand | Quantity | Processing | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Processing | Price | Quantity & Availability | |||||
11511851
|
Cytiva
SH30088.02 |
100 mL | None |
£139.00
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12399842
|
Cytiva
SH30088.02HI |
100 mL | Heat Inactivated |
£130.00
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16271102
|
Cytiva
SH30088.03IR25-40 |
500 mL | Irradiated, 25 to 40 doses |
£505.00
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11521851
|
Cytiva
|
500 mL | None |
£487.00
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12309852
|
Cytiva
SH30088.03HI |
500 mL | Heat Inactivated |
£555.00
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12339852
|
Cytiva
SH30088.03IR |
500 mL | Gamma Irradiated | N/A | ||||||
Description
- Support cell culture growth
- Standard grade
- Aseptically packaged in Class 100 cleanroom
Specifications
Gamma Globulin, Alkaline Phosphatase, Lactate Dehydrogenase, SGPT, SGOT, pH, Total Protein, Albumin, Blood Urea Nitrogen, Creatinine, Total Bilirubin, Sodium, Potassium, Calcium, Chloride, Inorganic Phosphorus, Osmolality, Iron, TIBC, IgG | |
United States | |
Sequential-filtered, 100 nm | |
Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus | |
USDA Inspected |
Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities | |
Sterile | |
Standard | |
No |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Cytiva HyClone™ Fetal Bovine Serum (U.S.), Standard