missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Fetal Clone I Serum
HyClone™ FetalClone™ I is a fetal bovine serum alternative optimised for the growth of hybridomas with comparable IgG levels to those found in FBS.
Brand: Cytiva SH30080.03IR
Code : 85
Additional Details : Weight : 0.50000kg
Specifications
Gamma Globulin, Alkaline Phosphatase, Lactate Dehydrogenase, SGPT, SGOT, pH, Total Protein, Albumin, Blood Urea Nitrogen, Creatinine, Total Bilirubin, Sodium, Potassium, Calcium, Chloride, Inorganic Phosphorus, Osmolality, Iron, TIBC, Percent Iron Saturation, Glucose, IgG | |
Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C. | |
United States | |
500 mL | |
Sequential-filtered, 100 nm, Irradiated | |
Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus | |
No | |
Typically less than 2 μg/mL | |
USDA Inspected |
Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities | |
FetalClone I Serum (U.S.) Irradiated | |
FBS Replacement Serum | |
Sterile | |
Irradiated | |
≤ 10 EU/mL | |
≤ 20 mg/dL | |
No |