missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ FetalClone™ II Serum (U.S.)
Optimized for the growth of CHO cells and derivatives.
Brand: Cytiva SH30066.02HI
95.20 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Code : 85
Additional Details : Weight : 0.20000kg
Description
Specifications
Gamma Globulin, Alkaline Phosphatase, Lactate Dehydrogenase, SGPT, SGOT, pH, Total Protein, Albumin, Blood Urea Nitrogen, Creatinine, Total Bilirubin, Sodium, Potassium, Calcium, Chloride, Inorganic Phosphorus, Osmolality, Iron, TIBC, Percent Iron Saturation, Glucose, IgG | |
United States | |
100 mL | |
Sequential-filtered, 100 nm | |
Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus | |
Yes | |
Typically less than 2 μg/mL | |
No |
Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2 to 8°C. | |
FBS Replacement Serum | |
Sterile | |
Heat Inactivated | |
≤ 10 EU/mL | |
≤ 20 mg/dL | |
HyClone™ | |
USDA Inspected |