missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ FetalClone™ II Serum (U.S.)

Optimized for the growth of CHO cells and derivatives.

Brand:  Cytiva SH30066.03IR

Code : 85

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 14850841

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

  • Economical alternative to FBS
  • Performance comparable with FBS
  • High lot-to-lot consistency
  • Low endotoxin and hemoglobin levels
  • demonstrated performance with a variety of cell lines, including hybridomas, CHO, BHK-21, NS0, MRC-5, and Vero cells
  • Specifications

    Specifications

    Gamma Globulin, Alkaline Phosphatase, Lactate Dehydrogenase, SGPT, SGOT, pH, Total Protein, Albumin, Blood Urea Nitrogen, Creatinine, Total Bilirubin, Sodium, Potassium, Calcium, Chloride, Inorganic Phosphorus, Osmolality, Iron, TIBC, Percent Iron Saturation, Glucose, IgG
    United States
    500 mL
    Sequential-filtered, 100 nm
    Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus
    No
    Typically less than 2 μg/mL
    No
    Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2 to 8°C.
    FBS Replacement Serum
    Sterile
    Irradiated
    ≤ 10 EU/mL
    ≤ 20 mg/dL
    HyClone™
    USDA Inspected
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents

    Documents

    Certificates
    Special Offers

    Special Offers