missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Hank's 1X Balanced Salt Solutions
Buffer cell culture growth with 1X balanced salt solutions
Brand: Cytiva SH30031.01
9.52 GBP valid until 2024-12-31
Use promo code "21631" to get your promotional price.
This item is not returnable.
View return policy
Description
- Used for washing and treating tissues and cells
Specifications
cGMP and ISO 9001:2000 Guidelines | |
1 X | |
Hanks' Balanced Salt Solution | |
Phenol Red | |
=1.0EU/mL | |
100 mL |
15° to 30°C | |
Liquid | |
0.1 μm Sterile-filtered | |
No Calcium, No Magnesium | |
7.0 to 7.4 |