missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Hank's 1X Balanced Salt Solutions
Buffer cell culture growth with 1X balanced salt solutions
£8.80 - £151.00
Specifications
Certifications/Compliance | cGMP and ISO 9001:2000 Guidelines |
---|---|
Content And Storage | 15° to 30°C |
Concentration | 1 X |
Form | Liquid |
Product Type | Hanks' Balanced Salt Solution |
Product Code | Brand | Quantity | With Additives | Without Additives | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | With Additives | Without Additives | Price | Quantity & Availability | |||
11546291
|
Cytiva
SH30588.02 |
1000 mL | No Calcium, No Magnesium, No Phenol Red |
£37.80
1000mL |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10553764
|
Cytiva
SH30031.FS |
6 x 500 mL | Phenol Red | No Calcium, No Magnesium |
£117.00
Pack of 6 |
Estimated Shipment: 11-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10739674
|
Cytiva
SH30268.01 |
500 mL | Calcium, Magnesium | No Phenol Red |
£15.15
500mL |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10041432
|
Cytiva
SH30031.03 |
1000 mL | Phenol Red | No Calcium, No Magnesium |
£22.30
1000mL |
Estimated Shipment: 11-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10532003
|
Cytiva
SH30031.02 |
500 mL | Phenol Red | No Calcium, No Magnesium |
£16.70
500mL |
Estimated Shipment: 09-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10617394
|
Cytiva
SH30588.01 |
500 mL | No Calcium, No Magnesium, No Phenol Red |
£18.85
500mL |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10309473
|
Cytiva
SH30031.01 |
100 mL | Phenol Red | No Calcium, No Magnesium |
£14.75
100mL |
Estimated Shipment: 20-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10225362
|
Cytiva
SH30030.02 |
500 mL | Calcium, Magnesium, Phenol Red |
£16.85
500mL |
Estimated Shipment: 06-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10452372
|
Cytiva
SH30268.02 |
1000 mL | Calcium, Magnesium | No Phenol Red |
£38.65
1000mL |
Estimated Shipment: 05-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10227993
|
Cytiva
SH30268.LS |
6 x 1000 mL | Calcium, Magnesium | No Phenol Red |
£151.00
Pack of 6 |
Estimated Shipment: 06-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10601993
|
Cytiva
SH30030.03 |
1000 mL | Phenol Red, Calcium, Magnesium |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
- Used for washing and treating tissues and cells
Specifications
cGMP and ISO 9001:2000 Guidelines | |
1 X | |
Hanks' Balanced Salt Solution | |
=1.0EU/mL |
15° to 30°C | |
Liquid | |
0.1 μm Sterile-filtered | |
7.0 to 7.4 |