missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Iron-Supplemented Calf Serum, U.S. origin
A cost-effective alternative to Fetal Bovine Serum (FBS). Contains additional iron to boost serum transferrin to physiological levels.
£14.75 - £137.00
Product Code | Brand | Processing | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Processing | Quantity | Price | Quantity & Availability | ||||
16201092
|
Cytiva
SH30072.03IR25-40 |
Irradiated, 25 to 40 doses | 500 mL |
£79.55
500mL |
Estimated Shipment: 20-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
16221092
|
Cytiva
SH30072.04IR25-40 |
Irradiated, 25 to 40 doses | 1000 mL |
£137.00
1 L |
Estimated Shipment: 20-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10761454
|
Cytiva
SH30072.02 |
None | 100 mL |
£14.75
100mL |
Estimated Shipment: 20-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
16231092
|
Cytiva
SH30072.04IR30-45 |
Irradiated, 30 to 45 doses | 1000 mL |
£137.00
1000mL |
Estimated Shipment: 20-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12319812
|
Cytiva
SH30072.02HI |
Heat Inactivate | 100 mL |
£30.00
100mL |
Estimated Shipment: 24-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15715533
|
Cytiva
SH30072.04HI |
Heat Inactivate | 1000 mL |
£137.00
1000mL |
Estimated Shipment: 20-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11551831
|
Cytiva
SH30072.03 |
None | 500 mL |
£98.70
500mL |
Estimated Shipment: 05-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12339812
|
Cytiva
SH30072.03HI |
Heat Inactivated | 500 mL |
£77.40
500mL |
Estimated Shipment: 14-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10339953
|
Cytiva
SH30072.04 |
None | 1000 mL |
£122.00
1000mL |
Estimated Shipment: 01-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12839447
|
Cytiva
SH30072.04IR |
Gamma Irradiated | 1000 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12349812
|
Cytiva
SH30072.03IR |
Gamma Irradiated | 500 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
HyClone Iron-Supplemented Bovine Calf Serum, US Origin is collected by venipuncture from formula-fed veal calves, which produce exceptionally high levels of transferrin. This serum is carefully supplemented with iron to load serum transferrin levels to physiological levels. After supplementation, the serum contains three to four times as much available iron and transferrin as fetal bovine serum (FBS) or equine serum.
Specifications
Sterile | |
Sequential-filtered, 100 nm |