missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Iron-Supplemented Newborn Bovine Calf Serum (New Zealand)
A cost-effective alternative to Fetal Bovine Serum (FBS).
Brand: Cytiva SH30626.03
Code : 85
Additional Details : Weight : 1.05000kg
Description
Specifications
Gamma Globulin, Alkaline Phosphatase, Lactate Dehydrogenase, SGPT, SGOT, pH, Total Protein, Albumin, Blood Urea Nitrogen, Creatinine, Total Bilirubin, Sodium, Potassium, Calcium, Chloride, Inorganic Phosphorus, Osmolality, Iron, TIBC, Percent Saturation, Glucose, IgG | |
New Zealand | |
1000 mL | |
Sequential-filtered, 100 nm | |
Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus | |
≤ 25 mg/dL | |
Yes |
Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2 to 8°C. | |
Newborn Calf Bovine Serum | |
Sterile | |
None | |
No | |
HyClone™ |