missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ L-Glutamine, Powder
For supplementing serum- and protein-free media
Brand: Cytiva SH30336.03
Additional Details : Weight : 0.50000kg
Description
- Support the growth of cells that have high energy demands
- Synthesize large amounts of proteins and nucleic acids
Specifications
L-Glutamine | |
Powder | |
Supplementing Serum- and Protein-Free Media | |
500 g |
Store from 15°C to 30°C | |
L-Glutamine | |
HyClone™ |