missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ L-Glutamine, Powder
For supplementing serum- and protein-free media
£31.93 - £239.03
Specifications
Amino Acids | L-Glutamine |
---|---|
Form | Powder |
Product Type | L-Glutamine |
For Use With (Application) | Supplementing Serum- and Protein-Free Media |
Product Line | HyClone™ |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17672075
|
Cytiva
SH30336.01 |
25 g |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
10628574
|
Cytiva
SH30336.02 |
100 g |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11541911
|
Cytiva
SH30336.03 |
500 g |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Support the growth of cells that have high energy demands
- Synthesize large amounts of proteins and nucleic acids
Specifications
L-Glutamine | |
L-Glutamine | |
HyClone™ |
Powder | |
Supplementing Serum- and Protein-Free Media |