Learn More
Cytiva HyClone™ Newborn Bovine Calf Serum
Supplement cell culture growth with New Zealand sourced Cytiva HyClone Newborn Bovine Calf Serum, an excellent and cost-effective alternative to fetal bovine serum (FBS).
Brand:  Cytiva SH30401.02IH25-40
 View more versions of this product
Product Code. 17682075
  • £173.00 / 1 L
Explore more special offers
Description
Description
  • Supplement cell culture growth with New Zealand sourced Cytiva HyClone Newborn Bovine Calf Serum, an excellent and cost-effective alternative to fetal bovine serum (FBS).
    • Sourced from animals that are typically less than 10 days old
    • Traceable
    • Conforms to USDA regulations
TRUSTED_SUSTAINABILITY
Specifications
Specifications
-10°C or Lower, Once Thawed Store at 2°C to 8°C
Newborn Calf Bovine Serum
1000 mL
New Zealand
25 to 40 Doses
Enhance Growth of Cells