missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ Newborn Bovine Calf Serum, Heat Inactivated

Augment cell culture growth with this cost-effective alternative to Fetal Bovine Serum (FBS)

Brand:  Cytiva SH30118.03HI

 View more versions of this product

Product Code. 12664237

  • £134.00 / 500mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Produced using careful collection and processing procedures, including venipuncture, to provide the highest quality product available
  • Ensures excellent culture results without fear of contamination
  • HyClone™ maintains complete control of the entire collection and production process for 100% traceability
  • Newborn bovine calf serum undergoes the same careful collection and processing procedures (including venipuncture) used for other bovine serum products
  • Sourced from the United States with complete traceability back to original source
  • Age of animal at time of collection less than 10 days old
  • Low in antibodies and high in growth factors
  • Virus panel testing according to 9 CFR 113.53
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Store at or below -10°C
United States
Newborn Calf Bovine Serum
Sterile
Heat Inactivated
≤ 50 EU/mL
Yes
HyClone™
<10 days
Liquid
500 mL
Sequential-filtered, 100 nm
500 mL
Augmenting Cell Culture Growth
≤ 25 mg/dL
Frozen
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cytiva HyClone™ Newborn Bovine Calf Serum, Heat Inactivated > 500 mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.