missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Newborn Bovine Calf Serum, New Zealand Origin
A cost-effective alternative to Fetal Bovine Serum (FBS).
Brand: Cytiva SH30401.02HI
Code : 85
Additional Details : Weight : 1.50000kg
Description
Specifications
Gamma Globulin,Alkaline Phosphatase,Lactate Dehydrogenase,SGPT,SGOT,pH,Total Protein,Albumin,Blood Urea Nitrogen,Creatinine,Total Bilirubin,Sodium,Potassium,Calcium,Chloride,Inorganic Phosphorus,Osmolality,Iron,TIBC,Percent Iron Saturation,Glucose,IgG | |
Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C | |
Sterile | |
Heat inactivated | |
≤ 25 EU/mL | |
Yes |
USDA Regulations | |
1000 mL | |
0.1 μm filtration | |
Bluetongue,Bovine Adenovirus,Bovine Parvovirus,Bovine RSV,BVDV,Cytopathogenic Agents,Hemadsorbing Agents,Rabies Virus,Reovirus | |
Yes | |
Yes |