missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Newborn Bovine Calf Serum, New Zealand Origin
A cost-effective alternative to Fetal Bovine Serum (FBS).
Brand: Cytiva SH30401.02IR30-45
135.85 GBP valid until 2024-06-28
Use promo code "21691" to get your promotional price.
Additional Details : Weight : 1.00000kg
Description
Specifications
Gamma Globulin,Alkaline Phosphatase,Lactate Dehydrogenase,SGPT,SGOT,pH,Total Protein,Albumin,Blood Urea Nitrogen,Creatinine,Total Bilirubin,Sodium,Potassium,Calcium,Chloride,Inorganic Phosphorus,Osmolality,Iron,TIBC,Percent Iron Saturation,Glucose,IgG | |
Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C | |
Sterile | |
Irradiated, 30 to 45 doses | |
≤ 25 EU/mL | |
Yes |
USDA Regulations | |
1000 mL | |
0.1 μm filtration | |
Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus | |
No | |
Yes |