missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Phosphate Buffered Saline (PBS)
Buffer cell culture growth with PBS
Brand: Cytiva SH30256.02
21.96 GBP valid until 2024-07-31
Use promo code "21860" to get your promotional price.
Code : 83
Additional Details : Weight : 1.00000kg
Description
- QC-tested solution
Specifications
cGMP and ISO 9001:2000 Guidelines | |
1 X | |
Phosphate Buffered Saline (PBS) | |
No Calcium, No Magnesium, No Phenol Red | |
7.0 to 7.2 |
15° to 30°C | |
Liquid | |
0.1 μm Sterile-filtered | |
=1.0EU/mL | |
1000 mL |