missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Phosphate Buffered Saline (PBS)
Buffer cell culture growth with PBS
£16.05 - £141.00
Specifications
Certifications/Compliance | cGMP and ISO 9001:2000 Guidelines |
---|---|
Content And Storage | 15° to 30°C |
Form | Liquid |
Product Type | Phosphate Buffered Saline (PBS) |
Sterilization Method | 0.1 μm Sterile-filtered |
Product Code | Brand | Quantity | Concentration | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Concentration | Price | Quantity & Availability | ||||
10572193
|
Cytiva
SH30256.02 |
1000 mL | 1 X |
£25.05
1000mL |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10462372
|
Cytiva
SH30256.01 |
500 mL | 1 X |
£16.05
500mL |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10793004
|
Cytiva
SH30256.LS |
6 x 1000 mL | 1 X |
£141.00
Pack of 6 |
Estimated Shipment: 04-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10294922
|
Cytiva
SH30258.01 |
500 mL | 10 X |
£35.70
500mL |
Estimated Shipment: 16-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10356662
|
Cytiva
SH30258.02 |
1000 mL | 10 X |
£43.25
1000mL |
Estimated Shipment: 02-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10126473
|
Cytiva
SH30256.FS |
6 x 500 mL | 1 X |
£88.05
Pack of 6 |
Estimated Shipment: 07-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
- QC-tested solution
Specifications
cGMP and ISO 9001:2000 Guidelines | |
Liquid | |
0.1 μm Sterile-filtered | |
=1.0EU/mL |
15° to 30°C | |
Phosphate Buffered Saline (PBS) | |
No Calcium, No Magnesium, No Phenol Red | |
7.0 to 7.2 |