missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ SFM4CHO™ Media
Designed for high performance in a variety of culture vessels, including bioreactors. HyClone™ SFM4CHO™ Media is a protein-free formulation that contains no components of bovine origin.
Brand: Cytiva SH30518.01
152.95 GBP valid until 2024-12-31
Use promo code "21631" to get your promotional price.
This item is not returnable.
View return policy
Description
- Formulated using proprietary lipid complexing process for enhanced stability and growth promotion of various CHO cell lines
Specifications
cGMP | |
SFM4CHO media | |
Non-sterile | |
5 L |
2° to 8°C, away from light and moisture | |
Powder | |
No Glutamine, No Sodium Bicarbonate, No Poloxamer 188, No Phenol Red, No HEPES |