Learn More
Cytiva HyClone™ SFM4CHO™ Media
Designed for high performance in a variety of culture vessels, including bioreactors. HyClone™ SFM4CHO™ Media is a protein-free formulation that contains no components of bovine origin.
£26.50 - £161.00
Product Code | Brand | Quantity | Form | With Additives | Without Additives | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Form | With Additives | Without Additives | Price | Quantity & Availability | ||
10483522
|
Cytiva
SH30549.02 |
1000 mL | Liquid | Glutamine, Sodium Bicarbonate, Poloxamer 188 | No Phenol Red, No HEPES |
£73.99
1000mL |
Estimated Shipment: 06-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
10580083
|
Cytiva
SH30518.01 |
5 L | Powder | No Glutamine, No Sodium Bicarbonate, No Poloxamer 188, No Phenol Red, No HEPES |
£161.00
5 L |
Estimated Shipment: 27-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10376852
|
Cytiva
SH30548.02 |
1000 mL | Liquid | Sodium Bicarbonate, Poloxamer 188 | No Glutamine, No Phenol Red, No HEPES |
£61.05
1000mL |
Estimated Shipment: 27-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
10631243
|
Cytiva
SH30549.01 |
500 mL | Liquid | Glutamine, Sodium Bicarbonate, Poloxamer 188 | No Phenol Red, No HEPES |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
Description
- Formulated using proprietary lipid complexing process for enhanced stability and growth promotion of various CHO cell lines
Specifications
cGMP | |
SFM4CHO media |