missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ SFM4CHO™ Media
Designed for high performance in a variety of culture vessels, including bioreactors. HyClone™ SFM4CHO™ Media is a protein-free formulation that contains no components of bovine origin.
Marca: Cytiva SH30549.01
Code : A0
Detalles adicionales : Weight : 0.50000kg
Descripción
- Formulated using proprietary lipid complexing process for enhanced stability and growth promotion of various CHO cell lines
Especificaciones
cGMP | |
SFM4CHO media | |
SFM4CHO Liquid Media | |
Glutamine, Sodium Bicarbonate, Poloxamer 188 | |
500 mL |
2° to 8°C, away from light | |
Liquid | |
0.2 μm Sterile-filtered | |
No Phenol Red, No HEPES |