missing translation for 'onlineSavingsMsg'
Learn More
compliance-icons

Cytiva HyClone™ Trypsin Protease

Product Code. 10217723
100 mL, 25.0 g porcine trypsin (1:250/L) in HBSS, No Calcium, No Magnesium, No EDTA, No Phenol Red, 100mL
Click to view available options
Quantity:
100 mL
500 mL
Formulation:
0.5 g porcine trypsin (1:250/L gamma irradiated) in HBSS with 0.2 g/L EDTA
2.5 g porcine trypsin (1:250/L gamma irradiated) in HBSS with 0.2 g/L EDTA
2.5 g porcine trypsin (1:250/L gamma irradiated) in HBSS with 1 g/L EDTA
25.0 g porcine trypsin (1:250/L) in HBSS
Without Additives:
No Calcium, No Magnesium
No Calcium, No Magnesium, No EDTA, No Phenol Red
Unit Size:
100mL
500mL
This item is not returnable. View return policy

Product Code. 10217723

Brand: Cytiva SV30037.01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Derived from porcine pancreas. HyClone™ Trypsin Protease is used to disaggregate adherent cells from substrate or surface to which they are attached.

  • Derived from porcine pancreas
  • Gamma irradiated prior to hydration and filling
  • Formulated without calcium and magnesium
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store frozen
Concentration 2.5% (10 X)
Form Liquid
Sterilization Method 0.1 ÎĽm Sterile-filtered
With Additives Trypsin, porcine, Hanks’ Balanced Salt solution (HBSS)
Without Additives No Calcium, No Magnesium, No EDTA, No Phenol Red
Formulation 25.0 g porcine trypsin (1:250/L) in HBSS
Quantity 100 mL
Source Porcine pancreas
compliance-icons
Product Identifier
  • Trypsin 2.5%(10X) Solution
Signal Word
  • Danger
Hazard Category
  • Respiratory sensitiser Category 1
Hazard Statement
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : trypsin
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cytiva HyClone™ Trypsin Protease >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.