Learn More
HYDRANAL™ - Composite 2, Reagent for volumetric one-component Karl Fischer titration (methanol free), Honeywell™ Fluka™
Marke: Honeywell Fluka 34806-500ML
76.52 GBP valid until 2024-12-31
Use promo code "21961" to get your promotional price.
Code : NEW
Weitere Details : CAS Number : 288-32-4 Weight : 0.55500kg
Spezifikation
90°C (194°F) | |
HYDRANAL™-Composite 2 | |
111-90-0,288-32-4,7446-09-5,7553-56-2,693-98-1,68007-08-9 |
500 mL | |
1.11g/mL |
Sicherheit und Handhabung
- HYDRANAL-Composite 2
Signalwort
- Danger
Gefahrenkategorie
- Reproductive toxicity Category 1B
- Specific target organ toxicity after repeated exposure Category 2
- Carcinogenicity Category 2
Gefahrenhinweis
- H351-Suspected of causing cancer.
- H360D-May damage the unborn child.
- H373-May cause damage to organs.
Sicherheitshinweis
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Ergänzende Informationen
- MIXTURE LIST-Contains : imidazole, sulphur dioxide, iodine, 2-Methylimidazole, 1H-Imidazole monohydriodide