missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HYDRANAL™ - Coulomat CG, Reagent for coulometric KF titration (catholyte solution), Honeywell Fluka™
£89.24 - £480.19
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15654500
|
Honeywell-Hydranal
34840-50ML |
10 x 5 mL Ampules |
£89.24
50mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15664500
|
Honeywell-Hydranal
34840-6X25ML |
6 x 25 mL | N/A | ||||||
15674500
|
Honeywell-Hydranal
34840-6X50ML |
1 Kit |
£480.19
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Specifications
14426-21-2, 67-56-1, 134227-25-1, 7446-09-5 | |
UN1230 |
Safety and Handling
P210-P280-P304 + P340 + P312-P305 + P351 + P338 + P310-P370 + P378-P403 + P235
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- HYDRANAL-Coulomat CG
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Acute toxicity Category 3
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity after single exposure Category 1
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H311-Toxic in contact with skin.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H331-Toxic if inhaled.
- H370-Causes damage to organs.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : methanol, diethanolamine hydrochloride,Ethanol, 2,2'-iminobis-, hydriodide, sulphur dioxide
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title