missing translation for 'onlineSavingsMsg'
Learn More
HYDRANAL™ - CRM Water Standard 1.0, Certified reference material for Karl Fischer titration (water content 1 mg/g = 0.1%), Honeywell Fluka™ Product Code.: 15655825
compliance-icons

HYDRANAL™ - CRM Water Standard 1.0, Certified reference material for Karl Fischer titration (water content 1 mg/g = 0.1%), Honeywell Fluka™

Product Code. 15655825 Shop All Honeywell Fluka Products
10 x 4 mL
Click to view available options
Quantity:
10 x 4 mL
This item is not returnable. View return policy

Product Code. 15655825

Brand: Honeywell-Hydranal 3442640ML

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Density 1.000 g/cm3
Flash Point 43°C
Quantity 10 x 4 mL
CAS 100-66-3, 108-32-7
CAS Min % ≥90.0000%, ≥1.0000%
CAS Max % <100.0000%, <10.0000%
Boiling Point 153°C to 155°C
Packaging Ampoule
Molecular Formula C7H8O
UN Number UN2222
InChI Key RDOXTESZEPMUJZ-UHFFFAOYSA-N
SMILES COC1=CC=CC=C1
IUPAC Name anisole
Molecular Weight (g/mol) 108.14
PubChem CID 7519
ChEBI CHEBI:16579
Grade Certified Reference Material
Assay effective water coulometric
Chemical Name or Material HYDRANAL™ - CRM Water Standard 1.0, Certified reference material for Karl Fischer titration (water content 1 mg/g = 0.1%)
Show More Show Less
compliance-icons
Product Identifier
  • HYDRANAL CRM Water Standard 1.0
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
Supplemental information
  • MIXTURE LIST-Contains : anisole, propylene carbonate
  • Multiple Manufacturers
    Use promotion code 24138 to get amazing savings on products for your analysis
    Promo Code: 24138

    Expires2025-04-30

    Days

    46

    Hours

    22

    Minutes

    6

    Seconds

    26
  • Multiple Manufacturers
    Shop now and save up to 97% on top-quality products for your lab needs.
    Promo Code: 25908

    Expires2025-03-31

    Days

    16

    Hours

    22

    Minutes

    6

    Seconds

    26
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
HYDRANAL™ - CRM Water Standard 1.0, Certified reference material for Karl Fischer titration (water content 1 mg/g = 0.1%), Honeywell Fluka™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.