Learn More
Honeywell Fluka HYDRANAL™ - NEXTGEN Coulomat A-FA, Honeywell Fluka™
Anolyte for coulometric Karl Fischer titration of ketones and Li-ion battery electrolytes, preferred for cells with diaphragm, acetonitrile-based, free of alcohols (free of CMR substances)
Brand: Honeywell Fluka 34471-500ML
Description
- HYDRANAL™ is the most trusted brand in Karl Fischer titration because of its dedication to quality, consistency and continued innovation.
- Comprehensive product portfolio with 60 available products
- 40 years of innovation – check our NEXTGEN reagents HERE
- Dedicated technical service and support
- More than 800 Application Notes available on our website
- View our NEXTGEN Coulomat A-FA standard operating procedure
Specifications
HYDRANAL™ - NEXTGEN Coulomat A-FA | |
0.906 g/cm3 | |
1 x 500 mL | |
5.5 | |
<20.00%, ≥20.00%, <70.00%, <10.00%, <10.00% | |
Liquid | |
Soluble in water |
Orange | |
12°C to 13°C | |
Confidential Stabilizer 2, 75-75-2, Confidential Accelerator, Confidential Stabilizer 1, 75-05-8 | |
≥60.00%, ≥2.00%, ≥10.00%, ≥2.00%, ≥10.00% | |
Glass Bottle | |
2924 |
Safety and Handling
- HYDRANAL NEXTGEN Coulomat A-FA
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Flammable liquid Category 2
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H290-May be corrosive to metals.
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H332-Harmful if inhaled.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : acetonitrile, methanesulphonic acid, 1-Ethylimidazole
Special Offers
-
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30
-
Multiple Manufacturers
Chromatography and Chemicals Promotion
Explore our new promotion and get up to 70% off some of our most popular products.Promo Code: 24127Expires: 2025-05-31
New Offer Save Up to 70%
Your input is important to us. Please complete this form to provide feedback related to the content on this product.