missing translation for 'onlineSavingsMsg'
Learn More

Honeywell Fluka HYDRANAL™ - NEXTGEN Coulomat A-FA, Honeywell Fluka™

Anolyte for coulometric Karl Fischer titration of ketones and Li-ion battery electrolytes, preferred for cells with diaphragm, acetonitrile-based, free of alcohols (free of CMR substances)

£260.04 - £1366.55

Specifications

Flash Point 12°C to 13°C
Chemical Name or Material HYDRANAL™ - NEXTGEN Coulomat A-FA
Color Orange
pH 5.5
Packaging Glass Bottle
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
17893080
View Documents
Honeywell Fluka
34471-500ML
1 x 500 mL Glass Bottle
£260.04
500mL
Please to purchase this item. Need a web account? Register with us today!
17504956
View Documents
Honeywell Fluka
34471-6X500ML
6 x 500 mL Glass Bottle
£1366.55
6 bottles
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • HYDRANAL™ is the most trusted brand in Karl Fischer titration because of its dedication to quality, consistency and continued innovation.
    • Comprehensive product portfolio with 60 available products
    • 40 years of innovation – check our NEXTGEN reagents HERE
    • Dedicated technical service and support
    • More than 800 Application Notes available on our website
    • View our NEXTGEN Coulomat A-FA standard operating procedure
Specifications

Specifications

12°C to 13°C
Orange
Glass Bottle
Confidential Stabilizer 2, 75-75-2, Confidential Accelerator, Confidential Stabilizer 1, 75-05-8
HYDRANAL™ - NEXTGEN Coulomat A-FA
5.5
Liquid
Soluble in water
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : 720

missing translation for 'storageNote' : Ambient

Product Identifier
  • HYDRANAL NEXTGEN Coulomat A-FA

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Flammable liquid Category 2
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H290-May be corrosive to metals.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H332-Harmful if inhaled.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : acetonitrile, methanesulphonic acid, 1-Ethylimidazole

SDS
Documents

Documents

Product Certifications