missing translation for 'onlineSavingsMsg'
Learn More

HYDRANAL™ - Titrant 5, Reagent for volumetric two-component Karl Fischer titration, Honeywell™ Fluka™

Brand:  Honeywell-Hydranal 34801-2.5L


 View more versions of this product

Product Code. 15644420

  • £137.79 / 2.50 L

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

TRUSTED_SUSTAINABILITY
Specifications

Specifications

HYDRANAL™-Titrant 5
11°C (51.8°F)
UN1230
0.85g/mL
2.5 L
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • HYDRANAL Titrant 5

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 3
  • Specific target organ toxicity after single exposure Category 1
  • Specific target organ toxicity after repeated exposure Category 2

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301 + H311 + H331-Toxic if swallowed, in contact with skin or if inhaled.
  • H370-Causes damage to organs.
  • H373-May cause damage to organs.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : methanol, iodine

SDS
Documents

Documents

Certificates
Special Offers

Special Offers